Learn More
MilliporeSigma™ UV Lamps and Assemblies for Water Purification Systems
Description
Control bacteria and oxidize organics
- Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Specifications
Specifications
| Product Type | UV Lamp |
| For Use With (Equipment) | Elix 20/35/70/100, RiOs 35/50/100/150/200 |
| Wavelength | 254 nm |