Learn More
MilliporeSigma™ UV Lamps and Assemblies for Water Purification Systems
Control bacteria and oxidize organics
Supplier: MilliporeSigma™ ZFRES00UV
Description
Control bacteria and oxidize organics
- Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Chemical Identifiers
| UV Lamp |
Specifications
| UV Lamp | |
| ASM Pure Water, Older SDS Reservoir | |
| 254 nm |