missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ RIPAb+™ SMN RIP Validated Antibody and Primer Set
Description
Includes negative control mouse IgG antibody and control primers specific for the cDNA of human U1snRNP.
Spécifications
Spécifications
| Applications | Immunocytochemistry, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Numéro d’acquisition du gène | NP_000335.1 |
| Symboles de gène | SMNT, SMN1, BCD541, Gemin-1, SMA, SMA1, SMA2, SMA3, SMA4, SMA™, SMN, SMNC |
| Espèces hôtes | Mouse |
| Immunogène | Histidine-tagged recombinant protein corresponding to human SMN. |
| Méthode de purification | Protein G purfied |
| Quantité | 10 Assays/Set (approx. 5μg of antibody per RIP) |
| Espèces cibles | Human, Mouse, Xenopus species |
| Contenu et stockage | -20°C for 12 months. |
| Afficher plus de résultats |