missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ hnRNPA1 (M9 Region) RIP Validated Antibody and Primer Set

Numéro de catalogue. 03181MI
Modifier l'affichage
Click to view available options
Quantité:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantité
03181MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 03181MI Fournisseur MilliporeSigma™ Code fournisseur 03181

Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.

This RIPAb+ hnRNPA1 (M9 Region) -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control mouse IgG antibody and control primers specific for the mRNA of human E2F3.

Specifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Numéro d’acquisition du gène NP_002127
Symboles de gène hnRNPA1, HNRNPA1, HNRPA1
Espèces hôtes Mouse
Immunogène Recombinant protein corresponding to human hnRNP A1.
Méthode de purification Protein G purfied
Quantité 10 Assays/Set (approx. 5μg of antibody per RIP)
Espèces cibles Human
Contenu et stockage -20°C for 12 months.
Type de produit RIPAb+ hnRNPA1 (M9 Region) RIP Validated Antibody and Primer Set
Forme Purified
Isotype IgG2b κ
Show More Show Less