missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ hnRNP U RIP Validated Antibody and Primer Set

Catalog No. 03206MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03206MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03206MI Supplier MilliporeSigma™ Supplier No. 03206

This RIPAb+ hnRNP U -RIP Validated Antibody and Primer Set conveniently includes the hnRNP U antibody and the specific control PCR primers.

Includes negative control normal mouse IgG antibody and control primers specific for the cDNA of human Ribosomal Protein S19.

Specifications

Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Gene Accession No. NP_004492.2
Gene Symbols pp120, hnrnpu, HNRPU, SAF-A, SAFA, U21.1, hnRNPU, p120
Host Species Mouse
Immunogen Recombinant protein corresponding to human hnRNP U.
Purification Method Protein G purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human
Content And Storage -20°C for 12 months.
Product Type RIPAb+ hnRNP U RIP Validated Antibody and Primer Set
Form Purified
Isotype IgG2b κ
Show More Show Less