missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ FXR1 RIP Validated Antibody and Primer Set

Catalog No. 03176MI
Change view
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
03176MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 03176MI Supplier MilliporeSigma™ Supplier No. 03176

This RIPAb+ FXR1 -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Includes negative control mouse IgG antibody and control primers specific for the cDNA of human JUN.

Specifications

Applications Immunocytochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Description Negative control mouse IgG antibody and control primers specific for the cDNA of human JUN
Gene Accession No. NP_001013456.1
Gene Symbols hFXR1, FXR1
Host Species Mouse
Immunogen Recombinant protein corresponding to human FXR1.
Purification Method Protein G
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human, Mouse
Content And Storage -20°C for 12 months.
Product Type RIPAb+ FXR1 RIP Validated Antibody and Primer Set
Form Purified
Isotype IgG2a κ
Show More Show Less