missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Ultrafiltration Membranes - YM
High-recovery membranes for stirred cells
$226.00 - $503.00
Specifications
| Filter Type | Adsorptive |
|---|---|
| Material (Membrane) | Regenerated Cellulose |
| MWCO | 100 kDa |
| Filter Code | YM-100 |
| For Use With (Application) | Protein purification, protein concentration, protein desalting |
| Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (Metric) | Price | Quantity | |||||
|
14422AM
|
MilliporeSigma™
14422AM |
4.45 cm |
Pack of 10 Each for $389.00
|
|
|||||
|
14432
|
MilliporeSigma™
14432 |
6.35 cm |
Pack of 10 Each for $473.00
|
|
|||||
|
14442AM
|
MilliporeSigma™
14442AM |
7.6 cm |
Pack of 10 Each for $503.00
|
|
|||||
|
14412
|
MilliporeSigma™
14412 |
2.5 cm |
Pack of 10 Each for $226.00
|
|
|||||
Specifications
| Adsorptive | |
| 100 kDa | |
| Protein purification, protein concentration, protein desalting | |
| Ultrafiltration Disc |
| Regenerated Cellulose | |
| YM-100 | |
| 10 Pack |