missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Ultrafiltration Membranes - YM
High-recovery membranes for stirred cells
Supplier: MilliporeSigma™ 14412
Specifications
| Adsorptive | |
| 100 kDa | |
| 4.1 cm2 | |
| 10 Pack | |
| 0.98 in. |
| Regenerated Cellulose | |
| YM-100 | |
| Protein purification, protein concentration, protein desalting | |
| Ultrafiltration Disc | |
| 2.5 cm |