missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Thick Absorbent Pad, 34 mm

Catalog No. AP30034P0
Change view
Click to view available options
Material:
Cellulose
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
AP30034P0 Cellulose
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AP30034P0 Supplier MilliporeSigma™ Supplier No. AP30034P0

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Cellulose support pads increase flow rate and throughput

Cellulose support pads are used to reinforce filters in Millipore's monitors for contamination analysis. When saturated with growth medium, they can also be used for culturing microorganisms on membranes.To increase flow rate and throughput, our woven mesh spacers can be placed between filters in a serial filtration to prevent the downstream screen filter from in.blinding in. the upstream filter pores. Features & Benefits•Avoid membrane damage during high pressure or fast flow conditions •Spacers enable you to combine multiple stages of filtration into a single process step, saving time Applications:Air Monitoring, Environmental Monitoring, Aerosol Contamination Monitoring

Specifications

Color White
Material Cellulose
Type Thick Absorbent Pad
For Use With (Application) Used to reinforce filters in monitors for contamination analysis, culturing microorganisms on membranes
Diameter (English) 1.33 in.
Diameter (Metric) 3.4 cm