missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Spectroquant™ Surfactants (cationic) Cell Test

Numéro de catalogue. M1017640001
Change view
Click to view available options
Type:
Surfactants (cationic)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Type
M1017640001 Surfactants (cationic)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. M1017640001 Fournisseur MilliporeSigma™ Code fournisseur 1.01764.0001
Il en reste null

Comprend

One bottle of reagent T-1K, 25 reaction cells, one sheet of round stickers for numbering the cells

Combines higher sensitivity with easier handling, to ensure optimal results.

  • Use with Spectroquant NOVA 30/60/400, Spectroquant Pharo 100/300 and Multy photometers.
  • Method: photometric
  • Range: 0.05 to 1.50mg/L CTAB
  • Reference Form: CTAB
  • Measures dissolved cationic surfactants of the quaternary ammonium compound, imidazoline, and amine type
  • Packaging: EPS box
  • Quantity: 25 Tests/Pk.

Spécifications

Type Surfactants (cationic)
For Use With (Equipment) Spectroquant NOVA 30/60/400, Spectroquant Pharo 100/300 and Multy photometers
Includes One bottle of reagent T-1K, 25 reaction cells, one sheet of round stickers for numbering the cells
Packaging EPS box
Range 0.05 to 1.50 mg/L
Storage Requirements Stable when stored closed at 15°C to 25°C
Test Method Photometric