Learn More
MilliporeSigma™ Simplicity™ Ultrapure Water System: Simpak 1 Kit
For use with Simplicity and Simplicity 185 Ultrapure Water Systems
Supplier: MilliporeSigma™ SIMPAKKR1
Description
For use with Simplicity and Simplicity 185 Ultrapure Water Systems
- For water pretreated by reverse osmosis, electrodeionization or distillation
- Includes one Purification Pack; one self-venting SimFilter that removes residual particulates and bacteria; and one 0.05μm Reservoir Vent Filter that protects pretreated water from airborne contaminants
Spécifications
| Elix, Reverse Osmosis, Distilled | |
| Simplicity™ 185 Ultrapure Water System | |
| SimPak 1 |
| 0.05 μm | |
| One Simpak one purification pack, one reservoir vent filter and one 0.05μm Simfilter final filter |