Learn More
MilliporeSigma™ SeQuant™ ZIC™-pHILIC Polymeric Analytical PEEK Columns
PEEK coated HPLC column
Supplier: MilliporeSigma™ 1504640001
Description
The polymeric SeQuant™ ZIC™-pHILIC columns is designed for particularly difficult separations of polar and hydrophilic compounds. The high-performance, truly zwitterionic stationary phase is attached to porous polymer beads and tolerates pH from 2-11.Specifications
| 4.6 mm | |
| 100 mm | |
| Column |