Learn More
MilliporeSigma™ RoClean™ Cleaning Pouches for Water Purification Systems
Catalog No. ZWBASE012
Change view
Click to view available options
Type:
Base Cleaning Pouch
Catalog No. Type
ZWBASE012 Base Cleaning Pouch
1 options
Catalog No. ZWBASE012 Supplier MilliporeSigma™ Supplier No. ZWBASE012
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Keep your purification system clean with easy-to-use RoClean pouches.
  • For cleaning RO membrane in Elix™, Milli-RO™, Milli-RX™, and low-volume Millipore™ RiOs™ Water Purification Systems
  • Available in acidic (ROClean A) and basic (ROClean B) formulas
†For details, see Catalog 2004/05.
Specifications
Type Base Cleaning Pouch
For Use With (Equipment) Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system