missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ RIPAb+™ Ago3 RIP Validated Antibody and Primer Set

Numéro de catalogue. 03250MI
Modifier l'affichage
Click to view available options
Quantity:
10 Assays/Set (approx. 5μg of antibody per RIP)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
03250MI 10 Assays/Set (approx. 5μg of antibody per RIP)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. 03250MI Fournisseur MilliporeSigma™ Code fournisseur 03250

This RIPAb+ Ago3 -RIP Validated Antibody and Primer Set conveniently includes the antibody and the specific control PCR primers.

Ago3 (Eukaryotic translation initiation factor 2C 3) belongs to the argonaute family and contains a PAZ and a Piwi domain. Ago3 plays a role in the RNA interference (RNAi) pathway.

Spécifications

Applications Immunocytochemistry, Immunoprecipitation
Classification Monoclonal
Gene Accession No. NP_079128
Gene Symbols AGO3, EIF2C3
Host Species Mouse
Immunogen Recombinant protein corresponding to human Ago3
Purification Method Protein G purfied
Quantity 10 Assays/Set (approx. 5μg of antibody per RIP)
Target Species Human
Content And Storage -20°C for 12 months.
Product Type RIPAb+ Ago3 Antibody
Form Purified
Isotype IgG2a κ
Afficher plus de résultats Afficher moins de résultats