missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Reflectoquant™ Phosphate Test Strips

Catalog No. M1169780001
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1169780001 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1169780001 Supplier MilliporeSigma™ Supplier No. 1.16978.0001
Only null left

Includes

Reagent, if needed

Used in reflectometric test

  • Determines ortho-phosphate in soils, fertilizers and food after pretreatment

Range: 5 to 120mg/L

Order Info

Pack of 50 strips

Specifications

For Use With (Application) Used for the determination of ortho phosphate in soils, fertilizers and food after sample pretreatment
For Use With (Equipment) Reflectometric
Graduations 5 to 120mg/L
Includes 1 barcode strip, 1 bottle of reagent orthophosphate-1, 1 test vessel with stopper
Quantity 50 Tests
Range 5 to 120 mg/L
Storage Requirements 15°C to 25°C
Test Method Phosphormolybdenum blue
Type Phosphate Test Strip