Please login to your online account to display your discounted pricing

EMD Millipore Quant™ Test Strips


Easy-to-use test strips for field testing, preliminary semi-quantitative analysis or quality control

Manufacturer: emd millipore  1100440001

 View more versions of this product

Catalog No. M1100440001

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Field testing, preliminary semi-quantitative analysis or quality control
Includes 1-bottle CN-1 and CN-2 reagent, 1 red dosing spoon, 1 test vessel
Storage Requirements 15° to 25°C
Graduations 1-3-10-30mg/L
Quantity Each
Range 1 to 30ppm
Test Method Barbituric acid derivative
Volume (English) 0.006 gal.
Volume (Metric) 25mL
Product Type Cyanide Test