missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Q-Gard TL Polishing Packs for ELIX, RiOs™

Catalog No. QGARDTL01
Change view
Click to view available options
Polishing Pack; For ELIX, RiOs systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
QGARDTL01 Polishing Pack; For ELIX, RiOs systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QGARDTL01 Supplier MilliporeSigma™ Supplier No. QGARDTL01

missing translation for 'validMainframeMsg'

For ELIX, RiOs (30 to 200) systems

Contains virgin ion-exchange resins for Elix

Specifications

Purification Method Ion Exchange, Depth Filtration
For Use With (Equipment) RiOs™ (30 to 200), Elix™ Clinical (35 to 70), Elix™ Golfstream Clinical, Large Elix™ (20 to 100)
Model Q-Gard TL
Type Polishing Pack