Learn More
MilliporeSigma™ Purospher™ STAR RP-8 Endcapped Hibar™ RT HPLC Columns
Optimized for more polar compounds
Supplier:  MilliporeSigma™ 1514530001
 View more versions of this product
Catalog No. M1514530001
Only null left
Explore available promotions
Description
Description
  • Enhanced selectivity for positional isomers
  • Excellent peak symmetry for polar and basic compounds
  • Excellent peak symmetry for acidic, basic and chelating compounds
Specifications
Specifications
Column
150 mm
1.5 to 10.5
1/Pk.
L7
Silica
Reversed Phase
Reversed Phase
4.6 mm
5 μm
120 Å
330 m²/g
C8
17%
Yes
Column
Product Suggestions
Product Suggestions