missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ PureProteome™ Nickel Magnetic Beads

Catalog No. LSKMAGH02
Change view
Click to view available options
Quantity:
10 mL
2 x 1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LSKMAGH02 2 x 1 mL
LSKMAGH10 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LSKMAGH02 Supplier MilliporeSigma™ Supplier No. LSKMAGH02

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Offer yields comparable with other magnetic purification systems while offering high purities

  • No lysate clearing—crude cell lysates are processed by magnetic beads
  • Fast processing time—entire protocol is completed in less than one hour
  • High capacity—bind 5 to 15mg of protein per mL of settled beads
  • High affinity—binding can take place in buffer containing <5mM EDTA
  • Flexible scale—varying the bead quantity allows different protein yields
  • Flexible elution—native or denatured proteins
  • Automatable—hands-free screening process

Specifications

Binding Property 1 to 5.5mg/mL
Content And Storage 2°C to 8°C
Particle Size 10 μm
Quantity 2 x 1 mL
For Use With (Application) Purification of His-tagged recombinant Protein
Type Magnetic Affinity Bead