missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Polyclonal Mouse IgG
Description
Mouse IgG, whole molecule, purified from normal mouse serum. Purity: 95% by SDS-PAGE
Spécifications
Spécifications
| Applications | ELISA |
| Concentration | 25 mg/mL |
| Aperçu général | 25 mg/mL |
| Formule | Purified mouse polyclonal IgG in buffer containing 0.01M PBS, 0.15M sodium chloride, 0.1% sodium azide, pH 7.1. |
| Quantité | 10mg |
| Contenu et stockage | Stable for 1 year at from date of receipt. |
| Type de produit | IgG, Protein A purfied |