missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Nutrient Media

Catalog No. p-98065
Change view
Click to view available options
Media Type:
Brettanomyces Selective Broth
HPC and Total Count Broth
m-Endo Total Coliform Broth
M-Green Selective Broth
mFC Broth
mFC Fecal Broth
Pseudomonas Selective Broth
Standard Plate Broth
Standard Plate Broth w/ Indicator
Yeast/Mold Selective
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Media Type
MHA000P2E m-Endo Total Coliform Broth
MHA000P2M M-Green Selective Broth
MHA000P2F mFC Fecal Broth
MHA00BSM2 Brettanomyces Selective Broth
MHA000P2T Standard Plate Broth w/ Indicator
MHA000P2S HPC and Total Count Broth
MHA000P2P Pseudomonas Selective Broth
MHA00FCR2 mFC Broth
MHA00P2TT Standard Plate Broth
MHA00P2SM Yeast/Mold Selective
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Catalog No. MHA000P2E Supplier MilliporeSigma™ Supplier No. MHA000P2E

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Developed specifically for culturing microorganisms on membrane filters; select media type to match organism type and analytical test needs

  • Each dry powder lot for media preparation assayed for purity and compared to satisfactory lot
  • Each m-Endo broth for total coliform lot and m-FC broth for fecal coliform analysis lot certified in accordance with the Safe Drinking Water Act
  • Lot QC certificates included in each package as added assurance for critical procedures

Order Info

Nutrient media comes in liquid form, packaged in 2mL plastic ampules. Refrigerate as detailed on label.

Specifications

Color Pink
Content And Storage 2°C to 10°C
Form Liquid
Manufacturing Origin United States
Media Type m-Endo Total Coliform Broth
Product Type Microbiology Media
Target Organism Class Coliforms
For Use With (Application) Water testing for Coliforms
Packaging Type Ampule
Quantity 50 x 2 mL