missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™HTS 8-Well Strips with Immobilon™-P PVDF Membrane

Catalog No. M8IPS4510
Change view
Click to view available options
Product Type:
Microplate 8-Well Strip
Microplate Strip Support Frame
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
M8IPS4510 Microplate 8-Well Strip
M8IPFRAME Microplate Strip Support Frame
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M8IPS4510 Supplier MilliporeSigma™ Supplier No. M8IPS4510

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Twelve 8-well strips in a support frame. Additional support frames also available separately.

8-well strips enable processing of less than 96 samples at one time

  • For ELISpot and protein binding assays

Specifications

Format Strip Plate
Array 1 x 8
Color Clear
Product Type Microplate 8-Well Strip
Material ABS Plastic
No. of Wells 8
Sterility Sterile
For Use With (Application) ELIspot and protein binding assays
Quantity 10/Pk.