missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ Column Loaders

Catalog No. MACL09645
Change view
Click to view available options
Volume (Metric) Well:
25 μL
45 μL
100 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Well
MACL09645 45 μL
MACL09625 25 μL
MACL09600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MACL09645 Supplier MilliporeSigma™ Supplier No. MACL09645

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

No need to pipet slurries, gels, or resins; load all 96 wells simultaneously and uniformly

  • Constructed of aluminum with an abrasion-resistant, chemically inert, nonstick coating

Specifications

Format Standard
Color Black
Volume (Metric) Well 45 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Load 45μL resin volumes simultaneously into 96 wells of MultiScreen plate for resin or bead based assays, chromatography
Quantity 1/Pk.