missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MultiScreen™ 96-Well Assay Plates for ELISPOT Assays

Catalog No. S2EM004M99
Change view
Click to view available options
Product Type:
MultiScreen HTS
MultiScreen HTS-IP with Underdrain
MultiScreen IP
MultiScreen-HA
MultiScreen-IP with Underdrain
Material:
Acrylic
Hydrophobic PVDF
Polystyrene
Membrane:
Immobilon-IP
Immobilon-P
Mixed Cellulose Esters
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Material Membrane
S2EM004M99 MultiScreen IP Acrylic Immobilon-P
MAHAS4510 MultiScreen-HA Polystyrene Mixed Cellulose Esters
MAIPS4510 MultiScreen-IP with Underdrain Acrylic Immobilon-P
MSIPN4W50 MultiScreen HTS-IP with Underdrain Acrylic Immobilon-IP
MSIPS4W10 MultiScreen HTS Acrylic Immobilon-IP
351044 MultiScreen IP Hydrophobic PVDF Immobilon-P
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. S2EM004M99 Supplier MilliporeSigma™ Supplier No. S2EM004M99

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining

  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days

Specifications

Format Standard
Membrane Immobilon-P
Array 8 x 12
Pore Size 0.45 μm
Product Line MultiScreen
Color White
Product Type MultiScreen IP
Material Acrylic
No. of Wells 96
Sterility Sterile
Filtration Area 0.28 cm2
For Use With (Application) ELISPOT Assays
Quantity 10/Pk.
Show More Show Less