missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ MQuant™ Zinc Test Strips

Catalog No. M1179530001
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1179530001 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1179530001 Supplier MilliporeSigma™ Supplier No. 1.17953.0001
Only null left

Includes

Reagent

High-tech, fast and simple means for semiquantitative testing

Uses a dithizone method.

Small and easy to handle for on-site use
Fast results
Ideal for screening
PET film and low reagent content for easier disposal

Graduations: 0-4-10-20-50mg/L

Order Info

No. of tests: 100

Specifications

For Use With (Equipment) Colorimetric
Graduations 0-4-10-20-50mg/L
Includes 2 bottles reagent Zn-1, 1 test vessel
Quantity 100 Tests
Range 0 to 50 mg/L
Storage Requirements 15°C to 25°C
Test Method Dithizone
Type Zinc Test Strip