missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ MQuant™ Manganese Test Strips
Description
Includes
Reagent
Uses an oxidation/reduction method.
- Small and easy to handle for on-site use
- Fast results
- Ideal for screening
- PET film and low reagent content for easier disposal
Graduations: 2-5-20-50-100mg/L
Order Info
No. of tests: 100
Specifications
Specifications
| For Use With (Application) | Used for routine testing of water, industrial water and effluent |
| For Use With (Equipment) | Colorimetric |
| Graduations | 2-5-20-50-100mg/L |
| Includes | 1 bottle reagent Mn-1, 1 bottle reagent Mn-2 |
| Quantity | 100 Tests |
| Range | 2 to 100 mg/L |
| Storage Requirements | 15°C to 25°C |
| Test Method | Oxidation/Redox indicator |
| Type | Manganese Test Strip |