missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Milli-DI™ Water Purification System

Catalog No. ZFDI00001
Change view
Click to view available options
Contaminant Rejection (Microbiological):
Greater than99%
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Contaminant Rejection (Microbiological)
ZFDI00001 Greater than99%
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ZFDI00001 Supplier MilliporeSigma™ Supplier No. ZFDI00001

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

DI-PAK™ Deionization Cartridge (CPDI000L1), sold separately.

Battery-powered system produces 3 to 5L/day of Type II purified water directly from tap water

  • Supplies purified water at a flowrate of 0.125 gal./min. (0.5L/min.) attaining >1 megohm·cm resistivity at 25°C (77°F).
  • Requires DI-PAK Deionization Cartridge (CPDI000L1), sold separately.

Two-stage purification sequence

  • Activated carbon first removes chlorine and colloids
  • Ion exchange resins remove typical inorganic ions

 Optional Millipak-40 Final Filter connects directly to water supply or faucet to remove bacteria particles >22μm at point of use (requires Millipak-40 thread adapter, MDI0 2AD AP).

System Operation Features

  • LED indicates when it is time to change cartridges
  • Temperature-compensated resistivity cell activates a red/green LED to indicate water quality meets requirements Resistivity meter operates on one 9V battery
  • Minimum feedwater pressure: 14psi
  • Maximum feedwater pressure: 65psi
DI-PAK™ Deionization Cartridge (sold separately: CPDI000L1, CPDI0NJM1) 
  • Exclusive simple “plug-and-use” design Easily replaced in seconds with no risk of leakage
  • Available in two sizes
    • Large pack contains 0.7 gal. (3.2L) of ion-exchange resin capable of purifying up to 106 gal. (400L) of feedwater (400μS/cm and 0ppm CO2)
    • Medium pack contains 0.5 gal. (2.3L) of ion-exchange resin capable of purifying up to 79.3 gal. (300L) of feedwater (400μS/cm and 0ppm CO2)
  •  Applications: 

    • Water produced is suitable for chemical and electrochemical applications, as well as for making buffers, reagents and microbiological culture media
    • Can also be used as feedwater for Simplicity™ Ultrapure Water Systems, small autoclaves, and humidifiers, as well as general applications in analytical and bioscience laboratories

     

Specifications

Contaminant Rejection (Microbiological) Greater than99%
Bacterial Content review, move to another attribute?
Daily Output 1000 L
Feedwater Source Potable Tap Water
Flow Rate 0.5 to 0.7
Product Water Type Type 2
Resistivity >1
Total Organic Carbon <500
Type Low Volume Water Purification System
Feedwater Pressure 14 to 65 psi
Model Milli-DI
Particle Size >22μm
Temperature Range (English) 77°F
Temperature Range (Metric) 25°C
Dimensions (L x W x H) 6.75 x 10.62 x 16.87 in. (17 x 27 x 43 cm)
Length (English) 6.75 in.
Length (Metric) 17 cm
Width (English) 10.62 in.
Width (Metric) 27 cm
Height (English) 16.87 in.
Height (Metric) 43 cm
Depth (English) 6.7 in.
Depth (Metric) 17 cm
Weight (English) 12 lb.
Weight (Metric) 5.6 kg
Voltage 9 V
Show More Show Less