Learn More
MilliporeSigma™ LiChroCART™ Columns/Purospher™ RP-18 Endcapped Sorbents HPLC Cartridges, 5μm
Excellent peak symmetry with either basic or strongly acidic compounds
Supplier:  MilliporeSigma™ 1501680001
 View more versions of this product
Catalog No. M1501680001
Only null left
Explore available promotions
Description
Description
  • Robust and fast method development with simple, neutral eluents
  • Based upon a high purity, metal-free silica with a complete coverage of the silica surface with C18-ligands
  • High chemical stability
  • Key applications are the determination of various azo dyestuff amines and beta-blockers demonstrating the good resolution and high peak symmetry
Specifications
Specifications
Cartridge
125 mm
5 μm
90 Å
480 m²/g
C18
18%
Yes
Cartridge
4 mm
2175 psi (150 bar) to 2900 psi (200 bar)
2 to 8
1/Pk.
L1
Silica
Reversed Phase
Reversed Phase
Product Suggestions
Product Suggestions