Learn More
MilliporeSigma™ LiChroCART™ Column/LiChrospher™ 100 RP-18 Endcapped Sorbent HPLC Cartridges, 5μm
HPLC cartridge
Supplier:  MilliporeSigma™ 1509360001
 View more versions of this product
Catalog No. M1509360001
Only null left
Explore available promotions
Description
Description
LiChrospher™ RP-18 endcapped is a reliable and versatile traditionally produced spherical silica gel carrier withC-18 endcapped reversed phase properties. It is well suited for the separation of neutral, acidic and weak basic compounds. This sorbent is packed into a LiChroCART™ HPLC cartridge. The connection to the HPLC system is being accomplished by use of the manu-CART™ NT cartrige holder (1.51486).
Specifications
Specifications
Cartridge
25 mm
5 μm
100 Å
350 m²/g
C18
21.6%
Yes
Stainless Steel
4 mm
2175 psi (150 bar) to 2900 psi (200 bar)
2 to 7.5
3/Pk.
L1
Silica
Reversed Phase
Reversed Phase
Cartridge
Product Suggestions
Product Suggestions