Learn More
MilliporeSigma™ LiChroCART™ Column/LiChrospher™ 100 NH2 Sorbent HPLC Cartridges, 5μm
HPLC cartridge
Supplier:  MilliporeSigma™ 1509580001
 View more versions of this product
Catalog No. M1509580001
Only null left
Explore available promotions
Description
Description
LiChrospher™ NH2 posses polar and hydrophobic properties and can be used for normal phase, reversed phase and ion exchange chromatography. Typical applications are the separations of carbohydrates by reversed phase chromatography or the separsation of nucleotides by ion exhange chromatography. Thissorbent is packed into a LiChroCART™ HPLCcartridge. The connection to the HPLC system is being accomplished by use of the manu-CART™ NT cartrige holder (1.51486).
Specifications
Specifications
Guard Column
4 mm
5 μm
100 Å
350 m²/g
Amino (NH2)
4.6%
Reversed Phase, Normal Phase, Ion Exchange, HILIC
Cartridge
4 mm
2175 psi (150 bar) to 2900 psi (200 bar)
2 to 7.5
10/Pk.
L8
Silica
Mixed Mode
Stainless Steel
Product Suggestions
Product Suggestions