missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Human RANTES Recombinant Protein, Animal Free
Spécifications
Spécifications
| Nom | Human RANTES Recombinant Protein, Animal Free |
| Quantité | 20 μg |
| Sources | E. coli |
| Recombinant | Recombinant |
| Grade de qualité ou de pureté | Endotoxin free and greater than 98% pure by SDS-PAGE gel and HPLC analyses |