missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Human iPS Reprogramming Boost Supplement
Description
Small molecules targeting specific signaling pathways have been shown to enhance the efficiency and/or replace the transcription factors required for reprogramming. EMD Millipore has screened multiple chemical compounds and selected the compounds in this kit based on their effects on enhancement of reprogramming efficiency as assessed by the number and overall quality and flat 2D morphology of human iPS colonies generated.
Spécifications
Spécifications
| Content And Storage | Kit contains sufficient quantities to supplement over 250mL of medium. HDAC Inhibitor-A Supplement (1000X): (Mfr. No. CS204423) One (1) vial containing 300μL of the inhibitor in water. TGF-β RI Kinase Inhibitor VI Supplement (1000X): (Mfr. No. CS204420) One (1) vial containing 300μL of the inhibitor in high quality DMSO. Store at -20°C |
| For Use With (Application) | Enhancing efficiency of human iPS colony formation by at least ten fold |
| Quantity | 1 Kit |