Learn More
MilliporeSigma™ Histosec™ Pastilles (without DMSO) Embedding Agent for Histology
Catalog No. M1151619025
Change view
Click to view available options
Quantity:
25kg
4 x 2.5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1151619025 25kg
M1151612504 4 x 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M1151619025 Supplier MilliporeSigma™ Supplier No. 1.15161.9025
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
solidification point 56-58°C, embedding agent for histology
Histosec™ pastilles (without DMSO) is an embedding medium for histology of samples of human origin. It is a paraffin enriched with polymers, also available with the addition of DMSO (dimethyl sulfoxide). DMSO increases the rate of penetration of paraffin and provides additional preservation of the tissue structures. Its melting range is between 56 - 58 °C and, within this range any specimen can be processed without fear of damage irrespective of the method used.
Specifications
Product Type Pastilles
Quantity 25kg