Learn More
MilliporeSigma™ ESGRO Complete™ Embryonic Stem Cell Line
Catalog No. SFCMTI2
Change view
Click to view available options
Quantity:
2 vials
Catalog No. Quantity
SFCMTI2 2 vials
1 options
Catalog No. SFCMTI2 Supplier MilliporeSigma™ Supplier No. SFCMTI2
derived from murine strain C57/BL6J are selected for high targeting efficiency and proven germ line transmission.
Embryonic Stem Cell Lines derived from murine strain C57/BL6J selected for high targeting efficiency and proven germ line transmission. Cells are supplied as 2.5 x 10
Specifications
Content And Storage Place vials in the vapor phase of liquid nitrogen storage immediately upon receipt until it is convenient to proceed to subculture
For Use With (Application) Stem Cell Culture
Quantity 2 vials