missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Chemicon™ MitoLight™ Mitochondrial Apoptosis Detection Kit
Specifications
Specifications
| Content And Storage | -20°C |
| Description | Mitochondrial Apoptosis Detection Kit |
| Detection Method | Fluorescence |
| For Use With (Application) | For flow cytometry utilizes a lipophilic cation, termed as MitoLight, as a mitochondrial activity marker |
| Includes | 10X Incubation Buffer, MitoLight |
| Quantity | 100 Tests |