missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ Apoptosis Inducer Set

Catalog No. APT800
Change view
Click to view available options
Quantity:
1/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APT800 1/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APT800 Supplier MilliporeSigma™ Supplier No. APT800

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Activity Assay

Specifications

Content And Storage -20°C
Description Apoptosis Inducer Set
For Use With (Application) Activity Assay
Includes Actinomycin D, Camptothecin, Cycloheximide, Dexamethasone and Etoposide
Quantity 1/Pk.