missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Chemicon™ ApopNexin Annexin V FITC Apoptosis Kit

Catalog No. APT750
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
APT750 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. APT750 Supplier MilliporeSigma™ Supplier No. APT750

missing translation for 'validMainframeMsg'

For flow cytometry to detect a specific biochemical change in the cell surface membrane that is considered to be a signature event of early apoptosis

Specifications

Content And Storage 2°C to 8°C
Description ApopNexin Annexin V FITC Apoptosis Kit
Detection Method Fluorescence
For Use With (Application) For flow cytometry to detect a specific biochemical change in the cell surface membrane that is considered to be a signature event of early apoptosis
Includes Propidium Iodide, ApopNexin™ FITC, Binding Buffer
Quantity 100 Tests