missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Calbiochem™ Avidin, Egg White

Catalog No. 18972510MG
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18972510MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18972510MG Supplier MilliporeSigma™ Supplier No. 18972510MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Glycoprotein containing four identical subunits

  • Form: White lyophilized solid
  • Molar Mass: 66,000
  • Biological Activity: ≥10 units/mg protein
  • Soluble in dH2O and dilute buffer
  • Storage: Store at -20°C

Specifications

Quantity 10 mg
Content And Storage -20°C