missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EmbryoMax™ PMEF, Neo Resistant, Strain FVB, Not Mytomycin Treated, Passage 1

Catalog No. PMEFNLP1
Change view
Click to view available options
Cell Type:
Primary MEF
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Cell Type
PMEFNLP1 Primary MEF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PMEFNLP1 Supplier MilliporeSigma™ Supplier No. PMEFNLP1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For stem cell culture applications

Specifications

Content And Storage -80°C
Cell Type Primary MEF
For Use With (Application) Stem cell culture
Quantity 1 vial

For Research Use Only