missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ EmbryoMax™ Embryonic Stem Cell Line, Strain C57/BL6, Passage 11

Catalog No. CMTI2
Change view
Click to view available options
Quantity:
2 vials (2.5 x 10^6/Ea.)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CMTI2 2 vials (2.5 x 10^6/Ea.)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CMTI2 Supplier MilliporeSigma™ Supplier No. CMTI2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For high targeting efficiency and germ line transmission

Specifications

Content And Storage Place vials in the vapor phase of liquid nitrogen storage immediately upon receipt until it is convenient to proceed to subculture.
Cell Type Embryonic Stem Cells (Strain C57/BL6)
For Use With (Application) Stem cell culture
Quantity 2 vials (2.5 x 10^6/Ea.)

For Research Use Only