missing translation for 'onlineSavingsMsg'
Learn More

Elma™ Elmasonic S50R Ultrasonic cleaner

Catalog No. 08000132
Change view
Click to view available options
Elmasonic S50R Ultrasonic cleaner
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
08000132 Elmasonic S50R Ultrasonic cleaner
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 08000132 Supplier Elma™ Supplier No. 104 9548

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Optimized for sieve cleaning, liquid degassing, and sample preparation. Tovatech Elmasonic S50R Ultrasonic cleaner removes soluble and insoluble particles from fine mesh sieves.

  • Accommodates 8 in. and 4 in. diameter sieves
  • Operating modes: Sieve clean, Sample prep, or Degas
  • Sieve clean thoroughly removes all lodged contaminants
  • Sample prep mixes and dissolves samples and cleans instruments
  • Degas rapidly removes gasses from samples, HPLC solvents and cleaning solutions
  • Set timer up to 30 minutes or continuous with manual shutoff
  • Auto shutoff after 12 hours
  • Cavitation-resistant stainless steel tank for long service life
  • Easily cleaned, colorful Stainless steel exterior

Warranty

Two year warranty

Specifications

Timer 1 to 30 min. and continuous
Material Stainless Steel
Frequency 37 kHz
Voltage 115/120 V
Capacity (English) 1.4 gal.
Capacity (Metric) 5.29 L
Dimensions (D x W x H) Interior 9.4 x 9.4 x 5.1 in.
For Use With (Application) Removes Contaminants From Laboratory Sieves. Mix, Dissolves, Disperse, or Homogenize Samples, Degas Solvents. Cleans Parts and Instruments.
Includes Power cord, Instruction manual, Warranty card
Model S50R
Wattage 150 w
Warranty 2 Year Warranty
Show More Show Less