Learn More
Biomatik Corporation Phospho-MAP3K8 (Thr290) Colorimetric Cell-Based ELISA Kit
Numéro de catalogue. 502291223
Encompass
Phospho-MAP3K8 (Thr290) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MAP3K8 (Thr290) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41279. Gene ID: 1326. Target Synonyms: C-COT; COT proto-oncogene serine/threonine-protein kinase; Cancer Osaka thyroid oncogene; M3K8; MAP3K8; Mitogen-activated protein kinase kinase kinase 8; TPL-2; TPL2; Tpl2; Tumor progression locus 2; kinase Cot. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Numéro de catalogue. 502291223 Fournisseur Biomatik Corporation Code fournisseur EKA51015
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Phospho-MAP3K8 (Thr290) Colorimetric Cell-Based ELISA Kit for detecting Phospho-MAP3K8 (Thr290) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P41279. Gene ID: 1326. Target Synonyms: C-COT; COT proto-oncogene serine/threonine-protein kinase; Cancer Osaka thyroid oncogene; M3K8; MAP3K8; Mitogen-activated protein kinase kinase kinase 8; TPL-2; TPL2; Tpl2; Tumor progression locus 2; kinase Cot. Restrictions: For Research Use Only. Not for use in diagnostic procedures.