Learn More
Biomatik Corporation Phospho-VAV2 (Tyr142) Colorimetric Cell-Based ELISA Kit
Numéro de catalogue. 502291542
Encompass
Phospho-VAV2 (Tyr142) Colorimetric Cell-Based ELISA Kit for detecting Phospho-VAV2 (Tyr142) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52735. Gene ID: 7410. Target Synonyms: Vav-2 protein; Guanine nucleotide exchange factor VAV2; VAV-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Numéro de catalogue. 502291542 Fournisseur Biomatik Corporation Code fournisseur EKA51384
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Phospho-VAV2 (Tyr142) Colorimetric Cell-Based ELISA Kit for detecting Phospho-VAV2 (Tyr142) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P52735. Gene ID: 7410. Target Synonyms: Vav-2 protein; Guanine nucleotide exchange factor VAV2; VAV-2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.