Learn More
Biomatik Corporation Phospho-Interferon- Receptor (Tyr457) Colorimetric Cell-Based ELISA Kit
Numéro de catalogue. 502291473
Encompass
Phospho-Interferon- Receptor (Tyr457) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Interferon- Receptor (Tyr457) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15260. Gene ID: 3459. Target Synonyms: CD119 antigen; CDw119; IFN-gamma-R1; INGR; INGR1; Interferon-gamma receptor alpha chain precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Numéro de catalogue. 502291473 Fournisseur Biomatik Corporation Code fournisseur EKA51307
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Phospho-Interferon- Receptor (Tyr457) Colorimetric Cell-Based ELISA Kit for detecting Phospho-Interferon- Receptor (Tyr457) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P15260. Gene ID: 3459. Target Synonyms: CD119 antigen; CDw119; IFN-gamma-R1; INGR; INGR1; Interferon-gamma receptor alpha chain precursor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.