Learn More
Biomatik Corporation Phospho-CaMK4 (Thr196/200) Colorimetric Cell-Based ELISA Kit
Numéro de catalogue. 502291434
Encompass
Phospho-CaMK4 (Thr196/200) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CaMK4 (Thr196/200) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16566. Gene ID: 814. Target Synonyms: Calcium/calmodulin-dependent protein kinase type IV catalytic chain; Calspermin; CAM kinase-GR; CaMK IV; CAMK4; CaMKIV; KCC4; kinase CaMK4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Numéro de catalogue. 502291434 Fournisseur Biomatik Corporation Code fournisseur EKA51262
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Phospho-CaMK4 (Thr196/200) Colorimetric Cell-Based ELISA Kit for detecting Phospho-CaMK4 (Thr196/200) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: Q16566. Gene ID: 814. Target Synonyms: Calcium/calmodulin-dependent protein kinase type IV catalytic chain; Calspermin; CAM kinase-GR; CaMK IV; CAMK4; CaMKIV; KCC4; kinase CaMK4. Restrictions: For Research Use Only. Not for use in diagnostic procedures.