Learn More
Biomatik Corporation Phospho-IGF1R (Tyr1165/Tyr1166) Colorimetric Cell-Based ELISA Kit
Numéro de catalogue. 502289069
Encompass
Phospho-IGF1R (Tyr1165/Tyr1166) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IGF1R (Tyr1165/Tyr1166) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08069. Gene ID: 3480. Target Synonyms: CD221; IGF1R; Insulin-like growth factor I receptor precursor; kinase IGF1R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Numéro de catalogue. 502289069 Fournisseur Biomatik Corporation Code fournisseur EKA51479
Pour de l'aide SVP utiliser la function de clavardage; nous envoyer un couriel à help@thermofisher.com ou appelez le service à la clientèle au 1-800-234-7437.
Phospho-IGF1R (Tyr1165/Tyr1166) Colorimetric Cell-Based ELISA Kit for detecting Phospho-IGF1R (Tyr1165/Tyr1166) in researchers plate their cell line of choice. Species Reactivity: Human (Homo sapiens), Mouse (Mus musculus), Rat (Rattus norvegicus). Detection Range: Qualitative. Sensitivity: At least 5000 cells/well. Format: 96-Well Plate. Assay Type: Cell-Based. Detection Method: Colorimetric. Assay Time: 4h, 30min. Accession Number: P08069. Gene ID: 3480. Target Synonyms: CD221; IGF1R; Insulin-like growth factor I receptor precursor; kinase IGF1R. Restrictions: For Research Use Only. Not for use in diagnostic procedures.