missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eleutheroside B, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Food and Beverages
Specifications
Specifications
| Packaging | Bottomless Glass Bottle with Inserted Fused Cone |
| CAS | 118-34-3 |
| Physical Form | Neat |
| Linear Formula | C17H24O9 |
| MDL Number | MFCD00016778 |
| Synonym | (E)-4-[-3-Hydroxy-1-propenyl]-2,6-dimethoxyphenyl-beta-D-glucopyranoside; Ilexanthin A; Ligustrin; Lilacin; Magnolenin; Methoxyconiferine; Sinapyl alcohol 4-O-glucoside; Syringin; Syringoside |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 372.37 |
| Percent Purity | ≥98.0% (HPLC) |
| Show More |