missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Electroporation Chambers: Petri Pulser Electrode

Catalog No. BTXPP352P
Change view
Click to view available options
Connection Type:
Banana Cables
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTXPP352P Banana Cables
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTXPP352P Supplier BTX™ Supplier No. 45 0130

Petri Pulser Electrode is an immersion electrode for adherent cells in situ.

  • Immersion electrode for adherent cells in situ
  • Works in 35 mm Petri dishes
  • Fits single well of 6-well plate

Specifications

Format Reusable electroporation applicator for 6-well plates or 35 mm petri dishe
For Use With (Application) Mammalian Cell Transfection, Gene Therapy, Protein or Drug Delivery, Plant/Yeast
For Use With (Equipment) Gemini X2, ECM 830, ECM 630, ECM 399, ECM 2001+ Generators
Material Gold-plated
Product Line BTX Electroporation
Product Type Electroporation accessory
Voltage 300 V
Description Petri Pulser
Field Type Homogenous
Length (English) Electrode 0.019 in.
Length (Metric) Electrode 0.5 mm
Pulse Length Range 1 μsec. to 35 msec.
Quantity 1 Ea.
Size (English) Gap 0.078 in.
Size (Metric) Gap 2 mm
Thickness 0.5 mm
Volume (Metric) 0.5 to 3 mL
Weight (English) 0.37 lb.
Weight (Metric) 0.17 kg
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Exponential Decay Wave, Square Wave
Show More Show Less