missing translation for 'onlineSavingsMsg'
Learn More

Electrolyte for Salt Analyzer Acetic Acid 1%, Sulfuric Acid 5%, with 0.01 g/L Sodium Chloride, Reagents

Catalog No. 01335408
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01335408 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01335408 Supplier Labchem Inc Supplier No. CE000900500A

missing translation for 'validMainframeMsg'

Electrolyte for Salt Analyzer Acetic Acid 1%, Sulfuric Acid 5%, with 0.01 g/L Sodium Chloride, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 100°C
Color Colorless
Packaging Poly Bottle
Quantity 500 mL
Solubility Completely miscible