missing translation for 'onlineSavingsMsg'
Learn More

Elaidic Acid 97.0+%, TCI America™

Catalog No. O00105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O00105G 5 g
O001025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O00105G Supplier TCI America Supplier No. O00105G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 112-79-8
Molecular Formula C18H34O2
Molecular Weight (g/mol) 282.468
MDL Number MFCD00063954
InChI Key ZQPPMHVWECSIRJ-MDZDMXLPSA-N
Synonym elaidic acid, trans-oleic acid, trans-9-octadecenoic acid, 9-octadecenoic acid, e-oleic acid, 9-octadecenoic acid, e, e-octadec-9-enoic acid, trans-elaidic acid, 9-octadecenoic acid, 9e, elaidinsaure
PubChem CID 637517
ChEBI CHEBI:27997
IUPAC Name (E)-octadec-9-enoic acid
SMILES CCCCCCCCC=CCCCCCCCC(=O)O

Specifications

Melting Point 44°C
Color White-Yellow
Boiling Point 288°C
Quantity 5 g
Formula Weight 282.47
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Elaidic Acid
EINECSNumber (2)-0609
RTECSNumber JX6125000
TSCA No
Recommended Storage Refrigerator